Skip to Content
Merck
All Photos(1)

Key Documents

MAB4065

Sigma-Aldrich

Anti-H-CAM Antibody, Pan, clone SFF-2

clone SFF-2, Chemicon®, from mouse

Synonym(s):

CD44, CD44std, CD44H, HCAM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 449.00

CHF 449.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 449.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 449.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

SFF-2, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG1κ

suitability

not suitable for Western blot

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CD44(960)

Specificity

Specific for human standard CD44. Does not distinguish between splice variants (exons v3v10).

Immunogen

Epitope: Pan

Application

Immunohistochemistry: 1 µg/mL; acetone fixed cryostat sections; 2 µg/mL following heat-induced epitope retrieval in 0.01 M citrate buffer, pH 6.0 (Banfalvi, 1999).
Immunofluorescence
Flow Cytometry: 1:20 as determined on U937 cells
Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Adhesion (CAMs)
This Anti-H-CAM Antibody, Pan, clone SFF-2 is validated for use in FC, IC, IH, IH(P) for the detection of H-CAM.

Linkage

Replaces: 04-1123

Physical form

Format: Purified
Liquid in PBS.

Storage and Stability

Prior to opening maintain at -20°C until expiration date. After opening maintain frozen at -20°C in undiluted aliquots until expiration date. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: Lymph node (lymphocytes), skin (keratinocytes), or tonsil.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Sook Lee et al.
Journal of controlled release : official journal of the Controlled Release Society, 205, 128-133 (2015-01-13)
Multipotent mesenchymal stem cells (MSCs) promise a therapeutic alternative for many debilitating and incurable diseases. However, one of the major limitations for the therapeutic application of human MSC (hMSC) is the lengthy ex vivo expansion time for preparing a sufficient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service