Skip to Content
Merck
All Photos(2)

Documents

MAB348B

Sigma-Aldrich

Anti-Alzheimer Precursor Protein A4 Antibody, clone 22C11, Biotin Conj. | MAB348B

clone 22C11, from mouse, biotin conjugate

Synonym(s):

Amyloid beta A4 protein, ABPP, APPI, APP, Alzheimer disease amyloid protein, Cerebral vascular amyloid peptide, CVAP, PreA4, Protease nexin-II, PN-II, N-APP, Soluble APP-alpha, S-APP-alpha, Soluble APP-beta, S-APP-beta, C99, Beta-amyloid protein 42, Beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22C11, monoclonal

species reactivity

rat, pig

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), mouse (based on 100% sequence homology), monkey (based on 100% sequence homology), human (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

Deposits of amyloid protein in senile plaques near nerve processes are found in the brains of aged humand and cases of Alzheimer′s Disease. The principle component of this extracellular amyloid is beta A4, a 4 kDa peptide derived from a larger amyloid precursor protein (APP), which is widely expressed in the brain and body. The creation, transport and function of these proteins is currently an active area of research.

Specificity

This antibody recognizes Alzheimer Precursor Protein A4 at the N-temrinus.

Immunogen

Epitope: N-terminus
Purified recombinant Alzheimer precursor A4 (pre A4695) fusion protein

Application

Evaluated by Immunohistochemistry in adult rat brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Alzheimer Precursor Protein A4 in adult rat brain tissue.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Quality

Evaluated by Immunocytochemistry in rat E18 primary cortex cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Alzheimer Precursor Protein A4 in rat E18 primary cortex cells.

Target description

Varies

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to Biotin in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat E18 primary cortex cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frances K Wiseman et al.
Brain : a journal of neurology, 141(8), 2457-2474 (2018-06-27)
Down syndrome, caused by trisomy of chromosome 21, is the single most common risk factor for early-onset Alzheimer's disease. Worldwide approximately 6 million people have Down syndrome, and all these individuals will develop the hallmark amyloid plaques and neurofibrillary tangles
Ahsan Habib et al.
Cell transplantation, 27(3), 438-455 (2018-03-22)
Alzheimer's disease (AD) is an age-related disorder that affects cognition. Our previous studies showed that the neuroprotective fragment of amyloid procurer protein (APP) metabolite, soluble APPα (sAPPα), interferes with β-site APP-cleaving enzyme 1 (BACE1, β-secretase) cleavage and reduces amyloid-β (Aβ)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service