Skip to Content
Merck
All Photos(2)

Key Documents

MAB2254

Sigma-Aldrich

Anti-Tropomyosin Antibody, clone 15D12.2

clone 15D12.2, from mouse

Synonym(s):

tropomyosin 1 (alpha), cardiomyopathy, hypertrophic 3, tropomyosin 1 alpha chain, HTM-alpha, Alpha-tropomyosin, chromosome 15 open reading frame 13, Tropomyosin-1, sarcomeric tropomyosin kappa, alpha tropomyosin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 414.00

CHF 414.00


Estimated to ship on04 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 414.00


Estimated to ship on04 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

15D12.2, monoclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TPM1(7168)

General description

Tropomyosins (Tms) are a family of >40 highly conserved protein isoforms resulting from alternative splicing of four different genes during development and that are expressed in numerous cell types. These proteins form coiled-coil heterodimers which bind to actin polymers along the α-helical grove. Tropomyosin’s function varies depending on cell type in which it is expressed. Within muscle, Tms play a role in the regulation of actin-myosin interplay for the purpose of mediating muscle contraction. In non-muscle cells, studies suggest a role in the stabilization of actin filaments, along with protecting actin filaments from the severing action of gelsolin and ADF/cofilin depolymerization. Tms also appear to have a role in the moderation of access to actin for other actin-binding proteins. Tropomyosin has an integral role in maintaining cardiovascular homeostasis. Down-regulation of Tms expression has been observed in tumor cells indicating a possible role for Tms as tumor suppressors.

Specificity

The antibody recognizes the entire exon 9d of Tropomyosin.

Immunogen

Epitope: Exon 9d
Linear peptide corresponding to the entire exon 9d of rat Tropomyosin.

Application

Detect Tropomyosin using this Anti-Tropomyosin Antibody, clone 15D12.2 validated for use in WB, IH.
Immunohistochemistry Analysis: 1:300 dilution from a representative lot detected Tropomyosin in human striated muscle tissue.
Research Category
Cell Structure

Metabolism
Research Sub Category
Cytoskeleton

Muscle Physiology

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 0.5 µg/mL of the antibody detected Tropomyosin in 10 µg of mouse brain tissue lysate.

Target description

~ 30-40 kDa observed MWs. Tm6: 40 kDa; Tm1: 38 kDa; Tm2: 36 kDa; Tm3: 34 kDa; Tm5: 30 kDa

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher Stephen Thom et al.
BMC biology, 18(1), 52-52 (2020-05-16)
Identifying causal variants and genes from human genetic studies of hematopoietic traits is important to enumerate basic regulatory mechanisms underlying these traits, and could ultimately augment translational efforts to generate platelets and/or red blood cells in vitro. To identify putative
Sophie Uzureau et al.
Cell reports, 30(11), 3821-3836 (2020-03-19)
The C-terminal variants G1 and G2 of apolipoprotein L1 (APOL1) confer human resistance to the sleeping sickness parasite Trypanosoma rhodesiense, but they also increase the risk of kidney disease. APOL1 and APOL3 are death-promoting proteins that are partially associated with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service