MAB1582
Anti-Chondroitin Sulfate Proteoglycan Antibody, carbohydrate epitope, clone Cat-316
ascites fluid, clone Cat-316, Chemicon®
Synonym(s):
CSPG
Select a Size
CHF 424.00
Estimated to ship on30 April 2025
Select a Size
About This Item
CHF 424.00
Estimated to ship on30 April 2025
Recommended Products
biological source
mouse
Quality Level
antibody form
ascites fluid
antibody product type
primary antibodies
clone
Cat-316, monoclonal
species reactivity
feline, rat
manufacturer/tradename
Chemicon®
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgM
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
rat ... Cspg4(81651)
Specificity
Immunogen
Application
Immunocytochemistry: 1:2,500 on primary cultures of neurons.
Immunoblot: 1:200,000
Immunoprecipitation
Optimal working dilutions must be determined by the end user.
Cell Structure
Integrins
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service