Skip to Content
Merck
All Photos(3)

Key Documents

MAB1581-I

Sigma-Aldrich

Anti-Aggrecan Core Protein Antibody, clone Cat-315

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 164.00
100 μG
CHF 415.00

CHF 164.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
CHF 164.00
100 μG
CHF 415.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 164.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

Cat-315, monoclonal

mol wt

calculated mol wt 235.2 kDa
observed mol wt ~260 kDa

species reactivity

rat

species reactivity (predicted by homology)

feline

packaging

antibody small pack of 100 μg

technique(s)

western blot: suitable

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

-10 to -25°C

target post-translational modification

unmodified

Gene Information

feline ... ACAN(101084542)

General description

Aggrecan Core Protein (UniProt: A0A5F5XE37; also known as Anti-Chondroitin Sulfate Proteoglycan core protein, CSPG1) is encoded by the ACAN gene (Gene ID: 101084542) in cat. Chondroitin sulfate proteoglycans (CSPGs) are a family of large extracellular matrix molecules, composed of a central core protein to which a varying number of glycosaminoglycan (GAG) side chains are attached. They are widely expressed in the mammalian central nervous system. CSPG1 consists of a protein core and a chondroitin sulfate side chain. CSPG1 lacks a transmembrane domain and the entire molecule is extracellular and, therefore, is considered a constituent of brain extracellular matrix. It is synthesized with a signal peptide (aa 1-16, which is subsequently cleaved off to generate the mature form. CSPG1 plays a role in neural development, plasticity, and post-injury response. CSPGs are involved in cell adhesion, cell growth, receptor binding, and cell migration. They are reported to inhibit axon regeneration after spinal cord injury and contribute to glial scar formation, which blocks new axons from growing into the injury site. During development CSPGs are reported to act as guidance cues for developing growth cones. In the adult, they stabilize synaptic connections and prevent aberrant axonal sprouting. Clone Cat-315 is shown to recognize a chondroitin sulfate proteoglycan (CSPG) expressed on the surface of subsets of neurons in many areas of the mammalian CNS. (Ref.: Siebert, JR., and Osterhout, DJ. (2011). J. Neurochem. 119(1); 186-188; Mathews, RT., et al. (2002). J. Neurosci. 22(17); 7536-7547; Lander, C., et al. (1998). J. Neurosci. 18(1); 174-183).

Specificity

Clone Cat-315 is a mouse monoclonal antibody that detects Aggrecan core protein (CSPG1). It detects an epitope on the extracellular region.

Immunogen

Feline brain proteoglycans.

Application

Quality Control Testing

Evaluated by Western Blotting in Rat brain tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Aggrecan Core Protein in Rat brain tissue lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Aggrecan Core Protein in cortex tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:150 dilution from a representative lot detected Aggrecan Core Protein in rat brain tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Aggrecan Core Protein in Rat e18 cortical cells.

Immunoprecipitation Analysis: A representative lot immunoprecipitated Aggrecan Core Protein in Immunoprecipitation applications (Lander, C., et. al. (1998). J Neurosci. 18(1):174-83).

Immunohistochemistry Applications: A representative lot detected Aggrecan Core Protein in Immunohistochemistry applications (Grant, E., et. al. (2016). Cereb Cortex. 26(3):1336-1348; Lander, C., et. al. (1998). J Neurosci. 18(1):174-83; Quattromani, M.J., et. al. (2018). Mol Neurobiol. 55(3):2196-2213).

Immunofluorescence Analysis: A representative lot detected Aggrecan Core Protein in Immunofluorescence applications (Quattromani, M.J., et. al. (2018). Mol Neurobiol. 55(3):2196-2213).

Western Blotting Analysis: A representative lot detected Aggrecan Core Protein in Western Blotting applications (Choi, B.R., et. al. (2020). Cell Rep. 31(5):107540; Quattromani, M.J., et. al. (2018). Mol Neurobiol. 55(3):2196-2213; Lander, C., et. al. (1998). J Neurosci. 18(1):174-83).

Immunocytochemistry Analysis: A representative lot detected Aggrecan Core Protein in Immunocytochemistry applications (Lander, C., et. al. (1998). J Neurosci. 18(1):174-83).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Aggrecan Core Protein, clone Cat-315, Cat. No. MAB1581-I, is a mouse monoclonal antibody that detects Aggrecan Core Protein and is used in Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgM in PBS without azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service