Skip to Content
Merck
All Photos(1)

Key Documents

MAB1399Z-I

Sigma-Aldrich

Anti-Integrin αX Antibody, clone N418 Antibody, azide free

clone N418, from hamster(Armenian)

Synonym(s):

Integrin alpha-X, CD11 antigen-like family member C, Leukocyte adhesion glycoprotein p150, 95 alpha chain, Leukocyte adhesion receptor p150, 95, CD11c

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 320.00

CHF 320.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 320.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 320.00


Estimated to ship on24 May 2025


Request a Bulk Order

biological source

hamster (Armenian)

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N418, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Itgax(16411)

General description

Integrin alpha-X (UniProt: Q9QXH4; also known as CD11 antigen-like family member C, Leukocyte adhesion glycoprotein p150,95 alpha chain, Leukocyte adhesion receptor p150,95, CD11c) is encoded by the Itgax gene (gene ID: 16411) in murine species. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. CD11c is a single-pass type I membrane protein. It has an extracellular domain (aa 20-1116), a transmembrane domain (aa 1117-1137), and a cytoplasmic domain (aa 1138-1169). It is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off in the mature form. Low levels of CD11c are expressed on human granulocytes and monocytes, and higher expression levels are observed on macrophages, dendritic cells and hairly cell leukemias. CD11c associates with beta-2 subunit in a non-covalent manner and then serves as a receptor for fibrinogen where it recognizes the sequence G-P-R. CD11c is shown to mediate cell-cell interaction during inflammatory responses and is important in monocyte adhesion and chemotaxis. Clone N418 is known to recognize an epitope on the p150 alpha chain. (Ref.: Metlay, JP et al. (1990). J. Exp. Med. 171(5); 1753-1771).

Specificity

Clone N418 is a mouse monoclonal antibody that detects integrin alpha-X in murine tissue.

Immunogen

Mouse spleen dendritic cells.

Application

Anti-Integrin X, clone N418, azide free, Cat. No. MAB1399Z-I, is an armenian hamster monoclonal antibody that detects Integrin alpha-X and has been tested for use in Immunohistochemistry, Flow Cytometry, and Immunoprecipitation.
Immunoprecipitation Analysis: A representative lot detected Integrin X in Immunoprecipitation applications (Metlay, J.P., et. al. (1990). J Exp Med. 171(5):1753-71).

Immunohistochemistry Analysis: A representative lot detected Integrin X in Immunohistochemistry applications (Tan, C.S.E., et. al. (2017). PLoS Pathog. 13(4):e1006311; Roland, C.L., et. al. (2009). Mol Cancer Ther. 8(7):1761-71; Metlay, J.P., et. al. (1990). J Exp Med. 171(5):1753-71).

Flow Cytometry Analysis: A representative lot detected Integrin X in Flow Cytometry applications (Metlay, J.P., et. al. (1990). J Exp Med. 171(5):1753-71; Granucci, F., et. al. (1997). J Immunol. 159(4):1794-99; Cervantes-Barragan, L., et. al. (2007). Blood. 109(3):1131-37).
Research Category
Inflammation & Immunology

Quality

Evaluated by Immunohistochemistry in mouse spleen frozen tissue sections.

Immunohistochemistry Analysis: A 1:25 dilution of this antibody detected Integrin X in mouse spleen frozen tissue sections.

Target description

129.15 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified armenian hamster monoclonal antibody IgG in PBS without preservatives.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service