Skip to Content
Merck
All Photos(1)

Key Documents

MAB10050

Sigma-Aldrich

Anti-CCT-beta Antibody, clone F39 P7 F11

clone F39 P7 F11, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 383.00

CHF 383.00


Estimated to ship on27 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 383.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 383.00


Estimated to ship on27 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F39 P7 F11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PCYT1B(9468)

General description

This gene encodes a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the gene described in this record have been observed but have not been thoroughly characterized.

Specificity

CCT-beta

Immunogen

Synthetic Peptide corresponding to the C-Terminus of Human CCT-beta

Application

Research Category
Protein Trafficking
Research Sub Category
Chaperones
This Anti-CCT-beta Antibody, clone F39 P7 F11 is validated for use in WB for the detection of CCT-beta.
Western Blot Analysis: 0.1 - 0.5 ug/mL of this antibody detects a 60 kDa band of CCT-beta in HeLa cell lysate.

Optimal antibody dilution must be determined by end-user.

Target description

60 kDa

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2aκ in buffer containing PBS and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Tested on Hela cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heba Ghozlan et al.
Frontiers in oncology, 11, 663877-663877 (2021-05-18)
Uncontrolled proliferation as a result of dysregulated cell cycling is one of the hallmarks of cancer. Therapeutically targeting pathways that control the cell cycle would improve patient outcomes. However, the development of drug resistance and a limited number of inhibitors
Anne E Showalter et al.
Scientific reports, 10(1), 798-798 (2020-01-23)
Chaperonin-containing TCP-1 (CCT or TRiC) is a multi-subunit complex that folds many of the proteins essential for cancer development. CCT is expressed in diverse cancers and could be an ideal therapeutic target if not for the fact that the complex

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service