Skip to Content
Merck
All Photos(1)

Key Documents

GR37

Sigma-Aldrich

Anti-Vitamin D Receptor (Ab-1) Rat mAb (9A7γE10.4)

liquid, clone 9A7γE10.4, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 383.00

CHF 383.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 383.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

CHF 383.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9A7γE10.4, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

chicken, hamster, mouse, porcine, rat, amphibian, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... VDR(7421)

General description

Purified rat monoclonal antibody generated by immunizing rats with the specified immunogen and fusing splenocytes with mouse myeloma p3 NS 1/Ag4-1 (NS1) mouse myeloma cells. Recognizes both the occupied and unoccupied forms of vitamin D receptor.
Recognizes both occupied and unoccupied vitamin D receptor in breast carcinomas expressing the vitamin D receptor protein.
This Anti-Vitamin D Receptor (Ab-1) Rat mAb (9A7γE10.4) is validated for use in Frozen Sections, Gel Shift, WB, IP, Paraffin Sections for the detection of Vitamin D Receptor (Ab-1).

Immunogen

purified, chick intestine vitamin D receptor

Application

Frozen Sections (1:50-1:100)

Gel Shift (see comments)

Immunoblotting (1:200-1:400)

Immunoprecipitation (10 µl/mg protein lysate)

Paraffin Sections (1:50-1:100, sample pre-treatment required, see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Positive Control
Breast carcinoma tissue (see comments)

Other Notes

Brown, A.J. 1999. Nephrol. Dial. Transplant.14, 11.
Fleet, J.C. 1999. Nutr. Rev.57 60.
Evans, S.R., et al. 1998. Clin. Cancer Res.4, 1591.
Friedrich, M., et al. 1998. J. Histochem. Cytochem.46, 1335.
Issa, L.L., et al. 1998. Inflamm. Res.47, 451.
Kivineva, M., et al. 1998. J. Steroid Biochem. Mol. Biol.66, 121.
Nangia, A.K., et al. 1998. J. Steroid Biochem. Mol. Biol.66, 241.
Does not cross-react with estrogen and glucocorticoid receptors. About 50% of breast tumors express vitamin D receptor protein. For gel shift assays, order antibody at 1 mg/ml; contact Customer Service for additional information. For paraffin sections pre-treatment in 2N HCl for 20 min at 37°C prior to staining. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Misaki Hirose et al.
Peritoneal dialysis international : journal of the International Society for Peritoneal Dialysis, 33(2), 132-142 (2012-10-04)
Vitamin D plays an important role in calcium homeostasis and is used to treat secondary hyperparathyroidism among dialysis patients. The biologic activity of vitamin D and its analogs is mediated by vitamin D receptor (VDR), which is distributed widely throughout
Soraya Gutierrez et al.
The Journal of biological chemistry, 279(42), 43581-43588 (2004-08-10)
Vitamin D receptor (VDR) and Runx2 are key regulators of tissue-specific gene transcription. Using the bone-related osteocalcin (OC) gene, we have previously shown that Runx2 is required for the extensive chromatin remodeling that accompanies gene activation. Here, we have addressed
José L Costa et al.
BMC genomics, 10, 499-499 (2009-10-30)
The active form of Vitamin D, 1,25-dihydroxyvitamin D3 (1,25D), has strong anti-proliferative effects, yet the molecular mechanisms underneath this effect remain unclear. In contrast, the molecular mechanism of 1,25D for the regulation of calcium homeostasis has principally been resolved, demonstrating
Jae Young So et al.
The Journal of steroid biochemistry and molecular biology, 148, 111-121 (2014-12-30)
Tumor-initiating cells (also known as cancer stem cells) are the subpopulation of cells shown to be responsible for tumor initiation, maintenance and recurrence. In breast cancer, CD44(+)/CD24(-/low) cells were identified as tumor-initiating cells. We previously reported that a Gemini vitamin
Jae Young So et al.
PloS one, 8(1), e54020-e54020 (2013-01-18)
CD44, a transmembrane glycoprotein, is a major receptor for extracellular proteins involved in invasion and metastasis of human cancers. We have previously demonstrated that the novel Gemini vitamin D analog BXL0124 [1α,25-dihydroxy-20R-21(3-hydroxy-3-deuteromethyl-4,4,4-trideuterobutyl)-23-yne-26,27-hexafluro-cholecalciferol] repressed CD44 expression in MCF10DCIS.com basal-like human breast

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service