Skip to Content
Merck
All Photos(2)

Key Documents

E9SU711

Millipore

NovaSeptum® GO Preloaded System with High Purity Bag, 9-port

sterile; β-irradiated

Synonym(s):

NovaSeptum® GO High Purity Bag, Sterile_Sampling_Container

Sign Into View Organizational & Contract Pricing

Select a Size

1000 ML
CHF 2’338.00
250 ML
CHF 2’338.00
100 ML
CHF 2’339.00
50 ML
CHF 2’339.00

CHF 2’338.00


Estimated to ship on01 June 2025



Select a Size

Change View
1000 ML
CHF 2’338.00
250 ML
CHF 2’338.00
100 ML
CHF 2’339.00
50 ML
CHF 2’339.00

About This Item

UNSPSC Code:
41104105

CHF 2’338.00


Estimated to ship on01 June 2025


material

316 stainless steel cannula
PureFlex polyethylene bag (fluid contact layer)
TPE tubing
polyester body
silicone septum (platinum-cured)

Quality Level

Agency

according to ISO 11137 (sterilization)
meets ISO 146441
146441

meets ISO 88 biological reactivity (all component materials)

sterility

sterile; β-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

-20-50 °C temp. range (-4-122 °F) (for 1000 mL)
-80-50 °C temp. range (-112-122 °F) (for 50 mL to 250 mL)
0.30 bar max. pressure (4.35 psi) at ±25 °C (for 1000 mL)
0.50 bar max. pressure (7.25 psi) at ±25 °C (for up to 250 mL)

impurities

<2.15 EU/device bacterial endotoxins (LAL test)

fitting

female Luer-Lok® outlet (3-piece)
male Luer-Lok® outlet (3-piece)
male Luer-Lok® outlet (3-piece, containing a male, female and an injection site)

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pathogen testing
pathogen testing
pharma/biopharma processes
pyrogen testing
sterile sampling
sterility testing
viral therapy

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Device configuration: Single Unit Sampling - Pre-loaded system
The NovaSeptum® GO pre-loaded disposable magazine is available with various standard configurations using NovaSeptum® GO high purity bags, autoclavable bags, bottles, transfer tubes or accurate volume (AV) syringe units. Other configurations are available upon request. For an optimal aseptic solution, use the NovaSeptum® GO pre-loaded disposable magazine together with a 1.5" or 2" NovAseptic® connector and NovaSeptum® GO pre-loaded starter kit (5/9 shot stainless steel base). Ideal for sampling from aseptic and sterile processes, the NovaSeptum® GO sterile sampling system provides consistently representative samples, safely and securely.

Application

Regular Sampling Sample (with or without protective case).
Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, and fermentation applications

Packaging

E9SU711-10050 (9x50 ml bags, 3 units per box)
E9SU711-10100 (9x100 ml bags, 3 units per box)
E9SU711-10250 (9x250 ml bags, 3 units per box)
E9SU711-11000 (9x1000 ml bags, 3 units per box)

Components

  • Septum: Platinum-cured silicone
  • Body: Polyester
  • Cannula: ASTM® 316 L Stainless steel
  • Fluid contact layer (bag): Polyethylene film (PureFlex film)
  • Tubing: Thermoplastic elastomer (TPE)
  • Outlet Tubing: 3-piece Luer-Lok®, containing a male, female and an injection site

Preparation Note

Units are integrity tested at regular intervals during manufacturing.
Aqueous extraction contains < 2.15 EU per device as determined using the Limulus Amebocyte Lysate (LAL) test.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany
PUREFLEX is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service