CX0396
Carbon Disulfide
Meets Reagent Specifications for testing USP/NF monographs OmniSolv®
Synonym(s):
Carbon Disulfide, Carbon Bisulfide
About This Item
Recommended Products
grade
reagent
Quality Level
Agency
meets USP/NF
vapor pressure
398 hPa ( 20 °C)
product line
OmniSolv®
Assay
≥99.9% (GC)
form
liquid
impurities
≤0.03% Water (H2O)
≤30 ppm Benzene (C6H6)
evapn. residue
≤5 ppm
color
APHA: ≤10
bp
46.5 °C/1013 hPa
mp
-111.6 °C
transition temp
flash point -30 °C
solubility
2.1 g/L
density
1.26 g/cm3 at 20 °C
UV cutoff
≤382 nm
UV absorption
λ: 382 nm Amax: ≤1.00
λ: 388 nm Amax: ≤0.50
λ: 403 nm Amax: ≤0.10
λ: 410 nm Amax: ≤0.05
λ: 500 nm Amax: ≤0.005
shipped in
ambient
storage temp.
room temp
SMILES string
S=C=S
InChI
1S/CS2/c2-1-3
InChI key
QGJOPFRUJISHPQ-UHFFFAOYSA-N
Related Categories
Application
Preparation Note
Analysis Note
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1
Target Organs
Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
-22.0 °F - closed cup
Flash Point(C)
-30 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service