Documents
BLSP5
Millipore
mPAGE® Macroporous Sponge, 5pk
replacement part for mPAGE® Mini Wet Transfer System
About This Item
UNSPSC Code:
41105305
NACRES:
NA.32
packaging
pkg of 5 packs
manufacturer/tradename
Millipore
technique(s)
immunoblotting: suitable
western blot: suitable
General description
The mPAGE® Macroporous Sponge, 5pk, is a replacement part for the mPAGE® Mini Wet Transfer System (MWTS).
The mPAGE® Mini Wet Transfer System (MWTS) accommodates two gel holder cassettes for electrophoretic transfer of proteins from mini gels to PVDF or nitrocellulose blotting membrane using mPAGE® Transfer Buffer or a compatible transfer buffer.
Compatibility
Compatible with the mPAGE® Mini Wet Transfer System.
Legal Information
MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany