ATAKEY001
M Air T Allen key
Tool to unscrew the groundplate of M Air T Air Sampler, e.g. to exchange the battery pack.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
M Air T™
input
sample type pharmaceutical(s)
application(s)
environmental
compatibility
for use with M Air T™ Portable Air Tester
shipped in
ambient
Legal Information
M AIR T is a trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service