Skip to Content
Merck
All Photos(1)

Key Documents

AP503F

Sigma-Aldrich

Goat Anti-Mouse IgG γ chain Antibody, FITC conjugate, Species Adsorbed

1 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
CHF 219.00

CHF 219.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
CHF 219.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

CHF 219.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgG is one of the most abundant protein in human serum. IgG is important for our defence against microorganisms and the molecules are produced by B-lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 10 11 variants.

Specificity

Reacts with heavy chain of mouse IgG1, IgG2a, IgG2b and IgG3. Absorbed for mouse IgM and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins.

Application

Detect Mouse IgG γ chain using this Goat anti-Mouse IgG γ chain Antibody, FITC conjugate, Species Adsorbed validated for use in ELISA & IF.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suman Sengupta et al.
Cell biology international, 37(5), 449-457 (2013-03-16)
Transforming growth factor-β (TGF-β) is a potent inhibitor of the growth of normal mammary epithelial cells, and has a pleiotropic, context-dependent, concentration-dependent action. We found attenuation of TGF-β signalling in mammary adenoma carcinoma cells. Phosphorylation at the linker site of
Miki Soda et al.
Scientific reports, 9(1), 1490-1490 (2019-02-08)
Human tissue-specific stem cells (hTSCs), found throughout the body, can differentiate into several lineages under appropriate conditions in vitro and in vivo. By transfecting terminally differentiated cells with reprogramming factors, we previously produced induced TSCs from the pancreas and hepatocytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service