Skip to Content
Merck
All Photos(2)

Key Documents

ABT98

Sigma-Aldrich

Anti-Kinectin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Kinectin, CG-1 antigen, Kinesin receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 352.00

CHF 352.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 352.00


Estimated to ship on20 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KTN1(3895)

General description

Organelle transport is facilitated by molecular motors, such as kinesin and dynein, which interact with microtubules to transport cargo in an ATP-dependent manner. The shuttling of organelles is also dependent on kinectin, a transmembrane ER protein that associates with kinesin and stimulates its ATPase activity. Kinesin-kinectin dynamics have also been linked to focal adhesion growth. In addition, kinectin has been shown to activate the RhoG GTPase pathway which involves Rac1 and Cdc42 signaling. Kinectin is expressed in testis, ovary, spleen, thymus, prostate, colon, and small intestine.

Specificity

Other homologies: Mouse (82% Mouse sequence homology) and Rat (79% sequence homology).
This antibody recognizes the lumenal region of Kinectin.

Immunogen

Epitope: Lumenal region
KLH-conjugated linear peptide corresponding to the lumenal region of human Kinectin.

Application

Detect Kinectin using this Anti-Kinectin Antibody validated for use in Western Blotting, ICC.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Kinectin in HeLa and A431 cells.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blot in human testis tissue lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Kinectin in 10 µg of human testis tissue lysate.

Target description

~158 kDa observed. Uniprot describes 3 isoforms at ~149, ~150, and ~156 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human testis tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service