Skip to Content
Merck
All Photos(2)

Key Documents

ABS538

Sigma-Aldrich

Anti-FABP1 Antibody

serum, from rabbit

Synonym(s):

Fatty acid-binding protein, liver, Fatty acid-binding protein 1, Liver-type fatty acid-binding protein, L-FABP, Squalene- and sterol-carrier protein, SCP, Z-protein, p14

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CHF 292.00

CHF 292.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
CHF 292.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 292.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

FABP1, also known as Fatty acid-binding protein, Liver Fatty acid-binding protein, Fatty acid-binding protein 1, Liver-type fatty acid-binding protein, L-FABP, Squalene- and sterol-carrier protein, SCP, Z-protein, or p14, and encoded by the gene FABP1/FABPL, is an important fatty acid binding and transport protein. FABP1 is produced in the liver and is part of a family of lipid carrier proteins. FABP1 facilitate the transfer of fatty acids between extra and intracellular membranes. FABP1 level in the blood is used as a biomarker for obesity and type II diabetes. Additionally, recent research indicates that FABP1 can modulate stellate cell activation and this may lead to diet-induced nonalcoholic fatty liver disease, thus FABP1 is a key focus of metabolic research.

Immunogen

Recombinant protein corresponding to rat FABP1.

Application

Anti-FABP1 Antibody detects level of FABP1 & has been published & validated for use in FABP1.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected FABP1 in wild type mouse liver cytosol and not in LFABP-1 -/- mouse liver cytosol.

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected FABP1 in 10 µg of human liver tissue lysate.

Target description

~ 14 kDa observed

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service