Skip to Content
Merck
All Photos(1)

Key Documents

ABS457

Sigma-Aldrich

Anti-ASAH2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

N-acylsphingosine amidohydrolase, Non-lysosomal ceramidase 2, HNAC1, Neutral ceramidase, Acylsphingosine deacylase 2, ASAH2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 286.00

CHF 286.00


Estimated to ship on10 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 286.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 286.00


Estimated to ship on10 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASAH2(56624)

General description

N-acylsphingosine amidohydrolase 2 (ASAH2), also known as neutral ceramidase (N-CDase, NCDase), is a key regulator of sphingolipid signaling metabolites. ASAH2 is known to hydrolyze the sphingolipid ceramide into sphingosine and free fatty acid at an optimal pH of 6.5-8.5. Additionally, ASAH2 is suggested to act as a repressor of apoptosis both by reducing C16-ceramide, thereby preventing ceramide-induced apoptosis, and generating sphingosine, a precursor of the antiapoptotic factor sphingosine 1-phosphate. ASAH2 is also suggested to be involved in the digestion of dietary sphingolipids in intestine tract. In this role, ASAH2 is implied to act as a key enzyme for the catabolism of dietary sphingolipids and consequently, regulating the levels of bioactive sphingolipid metabolites in the intestinal tract. ASAH2 is expressed primarily in the intestine. It is also expressed with higher levels in kidney, skeletal muscle and heart.

Immunogen

KLH-conjugated linear peptide corresponding to human ASAH2.

Application

Anti-ASAH2 Antibody is a highly specific rabbit polyclonal antibody, that targets ASAH2 & has been tested in western blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected ASAH2 in 15 µg of HEK293 cell lysate.

Target description

~70 kDa observed. Uniprot describes 2 isoforms at ~82 kDa and ~86 kDa Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service