Skip to Content
Merck
All Photos(2)

Key Documents

ABS436

Sigma-Aldrich

Anti-ROCK2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Rho-associated coiled-coil containing protein kinase 2, Rho-associated protein kinase 2, Rho-associated coiled-coil-containing protein kinase II, p164 ROCK-2, ROCK2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 232.00

CHF 232.00


Estimated to ship on19 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 232.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 232.00


Estimated to ship on19 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rock2(19878)

General description

Rho-associated protein kinase 2 (ROCK2), also known as Rho kinase 2, Rho-associated, coiled-coil-containing protein kinase II, and p164 ROCK-2, is a member of the AGC serine/threonine protein kinase family. ROCK2 is a key regulator of actin cytoskeleton and cell polarity. It is involved in regulation of actin cytoskeleton organization, smooth muscle contraction, malignant cell transformation, tumor invasion and metastasis, stress fiber and focal adhesion formation, neurite retraction, motility and cell adhesion. Many of the key roles of ROCK2 are attributed to its ability to phosphorylate ADD1, BRCA2, CNN1, EZR, DPYSL2, EP300, MSN, MYL9/MLC2, NPM1, RDX, PPP1R12A, VIM, SORL1 and IRF4. ROCK2 regulates positively the activation of p42/MAPK1-p44/MAPK3 and the regulation of p90RSK/RPS6KA1 during myogenic differentiation. Alternatively, ROCK2 is a negative regulator of VEGF-induced angiogenic endothelial cell activation. Recent studies have shown that ROCK2 inhibits keratinocyte terminal differentiation and may play an important role in the timely initiation of centrosome duplication. It also may regulate closure of the eyelids and ventral body wall through organization of actomyosin bundles. Additional evidence suggests that ROCK2 plays a critical role in the regulation of spine and synaptic properties in the hippocampus.

Immunogen

KLH-conjugated linear peptide corresponding to human ROCK2.

Application

Anti-ROCK2 Antibody is a highly specific rabbit polyclonal antibody, that targets Rho-associated protein kinase 2 & has been tested in western blotting & IHC.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected ROCK2 in mouse brain tissue.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected ROCK2 in 15 µg of mouse brain tissue lysate.

Target description

~160 kDa observed. Uncharacterized bands may appear in some lysate(s).

Linkage

Replaces: MABN1037

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ning Ai et al.
Experimental and therapeutic medicine, 18(2), 1493-1499 (2019-08-01)
MicroRNA-466 was recently characterized as a tumor suppressor with known biological function in prostate cancer. The aim of the current study was to investigate the possible involvement of microRNA-466 in hepatocellular carcinoma (HCC). The current study demonstrated that the expression
Esak Lee et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(41), e2308941120-e2308941120 (2023-10-02)
Impaired lymphatic drainage and lymphedema are major morbidities whose mechanisms have remained obscure. To study lymphatic drainage and its impairment, we engineered a microfluidic culture model of lymphatic vessels draining interstitial fluid. This lymphatic drainage-on-chip revealed that inflammatory cytokines that
Karthiga Santhana Kumar et al.
Cell reports, 23(13), 3798-3812 (2018-06-28)
The microenvironment shapes cell behavior and determines metastatic outcomes of tumors. We addressed how microenvironmental cues control tumor cell invasion in pediatric medulloblastoma (MB). We show that bFGF promotes MB tumor cell invasion through FGF receptor (FGFR) in vitro and that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service