ABS2208
Anti-GPR161
Synonym(s):
G-protein coupled receptor 161
About This Item
biological source
rabbit
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
mol wt
calculated mol wt 60.3 kDa
observed mol wt ~60. kDa
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
mouse
packaging
antibody small pack of 100
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
storage temp.
-10 to -25°C
Gene Information
mouse ... Gpr161(240888)
Specificity
Immunogen
Application
Evaluated by Western Blotting in lysate from HEK293T cell transiently overexpressing human GPR161
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected GPR161 in lysate from HEK293T cell transiently overexpressing human GPR161.
Tested Applications
Immunofluorescence Analysis: A representative lot detected GPR161 in Immunofluorescence application (Shimada, H., et. al. (2017). Cell Rep. 20(2):384-396).
Immunoprecipitation Analysis: A representative lot immunoprecipitated GPR161 in Immunoprecipitation application (Mukhopadhyay, S., et. al. (2013). Cell. 152(1-2):210-23).
Immunocytochemistry Analysis: A representative lot detected GPR161 in Immunocytochemistry application (Mukhopadhyay, S., et. al. (2013). Cell. 152(1-2):210-23).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service