ABS1026
Anti-Rab27B Antibody
serum, from rabbit
Synonym(s):
Ras-related protein Rab-27B, C25KG
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, rat, mouse
technique(s)
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... RAB27B(5874)
General description
Immunogen
Application
Signaling
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected Rab27B in rat parotid gland lysate (Imai, A., et al. (2004). Journal of Cell Science. 117:1945-1953).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Rab27B in flg-Rab27B (Imai, A., et al. (2004). Journal of Cell Science. 117:1945-1953).
Immunohistochemistry Analysis: A representative lot detected Rab27B in rat parotid acinar cells (Imai, A., et al. (2004). Journal of Cell Science. 117:1945-1953).
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Rab27B in 10 µg of mouse brain tissue lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service