ABN402
Anti-Spartin Antibody
serum, from rabbit
Synonym(s):
Homeobox protein cut-like 1, CCAAT displacement protein, CDP, Homeobox protein cux-1, Spartin
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, rat, human
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CUX1(1523)
General description
Immunogen
Application
Neuroscience
Developmental Neuroscience
Western Blotting Analysis: A representative lot detected Spartin in HeLa, NSC34, and PC12 cell lysates (Edwards, T. L., et al. (2009). Biochem J. 423(1):31-39.).
Immunoprecipitation Analysis: A representative lot immunoprecipitated Spartin in HeLa cell lysate (Edwards, T. L., et al. (2009). Biochem J. 423(1):31-39.).
Immunofluorescence Analyis: A representative lot detected Spartin in HeLa cells (Edwards, T. L., et al. (2009). Biochem J. 423(1):31-39.).
Quality
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Spartin in 10 µg of HEK293 cell lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Analysis Note
HEK293 cell lysate
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service