Skip to Content
Merck
All Photos(2)

Key Documents

ABN307

Sigma-Aldrich

Anti-LGI1 Antibody

from rabbit

Synonym(s):

Leucine-rich, glioma inactivated 1, epitempin, EPT

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 352.00

CHF 352.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 352.00


Estimated to ship on27 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LGI1(9211)

General description

The leucine-rich, glioma inactivated gene 1 (LGI1) was first identified as tumor suppressor gene for glioma and may play a role in other cancers. LGI1 is a member of a family of highly related proteins containing leucine-rich repeats (LRRs) which are very similar to other transmembrane signaling molecules and receptors. LGI1 serves as a ligand to ADAM22, a metalloprotease localized at the synapse. Mutations in LGI1 account for nearly half of autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms.

Specificity

This antibody recognizes only Isoform 1.

Immunogen

KLH-conjugated linear peptide corresponding to human LGI1.

Application

Detect the LGI1 protein using this Anti-LGI1 validated for use in WB & IHC.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience
Western Blot Analysis: 2 µg/mL from a representative lot detected LGI1 in 10 µg/mL of HEK293 cell lysate.

Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected LGI1 in mouse cerebellum and rat cortex tissues.

Quality

Evaluated by Western Blot in mouse brain tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected LGI1 in 10 µg/mL of mouse brain tissue lysate.

Target description

~64 kDa observed. Uniprot describes 3 isoforms at ~33 kDa, ~58 kDa, and ~64 kDa An uncharacterized band may appear at ~45 kDa in some lysates.

Physical form

Antigen Affinity Purified
Format: Purified
Purified rabbit polyclonal in buffer containing PBS with 0.02% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service