Skip to Content
Merck
All Photos(1)

Key Documents

ABN190

Sigma-Aldrich

Anti-CEA cell adhesion molecule 16 (Ceacam16) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Carcinoembryonic antigen-related cell adhesion molecule 16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Carcinoembryonic antigen-related cell adhesion molecule 16, also known as CEA cell adhesion molecule 16 (Ceacam16), is a novel adhesion protein that has been found in cochlear hair cells. It interacts with α-tectorin-interacting protein and may play a role in maintaining the tectorial membrane. Mutation of the Ceacam16 gene results in autosomal dominant nonsyndromic deafness (ADNSHL).

The previously assigned protein identifier B2RUD6 has been merged into E9QA28. Full details can be found on the UniProt database.

Immunogen

Linear peptide corresponding to mouse Ceacam16 monomers and dimers.

Application

Immunohistochemistry Analysis: A representative lot was used by an independent laboratory in mouse cochlear tissue. (Zheng, J., et al. (2011).PNAS. 108(10):4218–4223.)

Immunofluorescence Analysis: A representative lot was used by an independent laboratory in mouse cochlear tissue. (Zheng, J., et al. (2011).PNAS. 108(10):4218–4223.)
Research Category
Neuroscience
Research Sub Category
Sensory & PNS
This Anti-CEA cell adhesion molecule 16 (Ceacam16) Antibody is validated for use in Western Blotting, IHC, Immunofluorescence for the detection of CEA cell adhesion molecule 16 (Ceacam16).

Quality

Evaluated by Western Blot in mouse cochlear tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Ceacam16 in 10 µg of mouse cochlear tissue lysate.

Target description

~55-250 kDa observed. This protein has been shown to detect monomers and dimers. (Zheng, J., et al. (2011).PNAS. 108(10):4218–4223.)

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse cochlear tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service