Skip to Content
Merck
All Photos(1)

Key Documents

ABN1699

Sigma-Aldrich

Anti-Prickle1

from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
CHF 139.00
100 μG
CHF 299.00

CHF 139.00


Estimated to ship on06 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
CHF 139.00
100 μG
CHF 299.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 139.00


Estimated to ship on06 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Prickle-like protein 1(UniProt: Q3U5C7) is encoded by the Prickle1 (also known as Pk1) gene (Gene ID: 106042) in murine species. Prickle Prickle-like Protein 1 is involved in the planar cell polarity (PCP) pathway that controls convergent extension during gastrulation and neural tube closure. PCP pathway comprises of multiple integral membrane proteins ((Frizzled, Flamingo/Stan/Celsr and Vangl/ Strabismus) and membrane-associated proteins (Dishevelled, Inversin, and Prickle). Prickle-like protein 1 is synthesized with a propeptide (aa 830-832) that is removed in mature protein. Prickle1 is expressed in distinct cell populations of the central nervous system and is a contributor to neuronal morphogenesis. Both Pk1 and Pk2 genes share three LIM (Lin 11, Isl-1 and Mec-3) and one PET domain (Prickle, Espinas and Testin) at the N-terminus of the protein and a CAAX prenylation motif at the C-terminus. Pk1 is shown to be expressed in the cortical subplate and is considered as an important site for early neuronal maturation. In the retinal tissue Pk1 expression is specific to neurons and is highly enriched in Starburst amarcine cells. Loss of Pk1 gene has been linked to early embryonic lethality.

Specificity

This rabbit polyclonal antibody detects Prickle-like protein 1 in murine species.

Immunogen

His-tagged Recombinant Protein corresponding to the C-terminus of Mouse Prickle-like protein 1.

Application

Anti-Prickle1 , Cat. No. ABN1699, is a highly specific rabbit polyclonal antibody that targets Prickle-like protein 1 and has been tested in Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A representative lot detected Prickle1 in Western Blotting applications (Liu, C., et. al. (2014). Biol Open. 3(9):861-70).

Quality

Evaluated by Western Blotting in mouse retinal tissue lysate.
Western Blotting Analysis: 1 µg/mL of this antibody detected Prickle1 in mouse retinal tissue lysate.

Target description

~94 kDa observed; 94.13 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service