Skip to Content
Merck
All Photos(1)

Key Documents

ABN111

Sigma-Aldrich

Anti-RGS11 (CT) Antibody

from rabbit

Synonym(s):

Regulator of G-protein signaling 11, RGS11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 311.00

CHF 311.00


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 311.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 311.00


Estimated to ship on11 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RGS11(8786)

General description

Regulator of G-protein signaling 11 (RGS11) is a multidomain GTPase containing one each of DEP, RGS, and G-protein gamma domains. Like other members of the R7 RGS family, RGS11 uses these domains to form multisubunit complexes, which process the light responses of retinal cells. RGS11 interacts with the RGS9 anchor protein (R9AP) and the 5 G-protein beta subunit (Gbeta5) to form a trimeric complex that regulates the activity of the metabotropic glutamate receptor 6 (mGluR6) in ON-bipolar cells. The RGS11 complex hydrolyzes GTP and inactivates the G-protein Galphao, which is activated by mGluR6 in dark environments. By this mechanism, the RGS11 complex accelerates the light response in ON-bipolar retinal cells.

Specificity

This antibody recognizes the C-terminus of RGS11.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of human RGS11.

Application

Detect RGS11 (C-terminus) using this Anti-RGS11 (C-terminus) Antibody validated for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Evaluated by Western Blot in human placenta tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected RGS11 in 10 µg of human placenta tissue lysate.

Target description

~53 kDa observed

Physical form

Affinity purified
Format: Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human placenta tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service