Skip to Content
Merck
All Photos(2)

Key Documents

ABE434

Sigma-Aldrich

Anti-TIF1-alpha Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Transcription intermediary factor 1-alpha, E3 ubiquitin-protein ligase TRIM24, RING finger protein 82, Tripartite motif-containing protein 24

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 318.00

CHF 318.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 318.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 318.00


Estimated to ship on22 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rhesus macaque (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TRIM24(8805)

General description

Transcription intermediary factor 1-alpha (TIF1-alpha) is also called E3 ubiquitin-protein ligase TRIM24, RING finger protein 82, and Tripartite motif-containing protein 24. TIF1-alpha is a transcriptional coactivator that interacts with chromatin depending on histone H3 modifications, with the highest affinity for histone H3 that is both unmodified at ′Lys-4′ (H3K4me0) and acetylated at ′Lys-23′ (H3K23ac). TIF1-alpha has E3 protein-ubiquitin ligase activity, helps regulate cell proliferation and apoptosis and up-regulates transcription activation. Aberrant TIF1-alpha can cause thyroid papillary carcinoma (TPC) and is up-regulated in some breast cancers.

Specificity

Other homologies: Rat (93% sequence homology).
This antibody recognizes the C-terminus of the Nuclear Receptor Binding Site (NRBS) of TIF1-alpha.

Immunogen

Epitope: C-terminus of the Nuclear Receptor Binding Site (NRBS)
KLH-conjugated linear peptide corresponding to the C-terminus of the Nuclear Receptor Binding Site (NRBS) of human TIF1-alpha.

Application

Detect TIF1α using this rabbit polyclonal antibody, Anti-TIF1-alpha Antibody validated for use in western blotting & ICC.
Immunocytochemistry Analysis: A 1:50 dilution from a represenative lot detected TIF1-alpha in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: 0.1 µg/mL of this antibody detected TIF1-alpha in 10 µg of HeLa cell lysate.

Target description

~140 kDa observed. Uniprot descrives 2 isoforms at ~117 kDa and ~113 kDa due to alternative splicing. This protein has been observed at ~140 kDa (Gunawardena, H., et al. (2008). Rheumatology (Oxford). 47(3):324-328.).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service