Skip to Content
Merck
All Photos(4)

Key Documents

ABE213

Sigma-Aldrich

Anti-monomethyl-Histone H3 (Lys79) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Histone H3.1t, H3/g

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CHF 434.00

CHF 434.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
CHF 434.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 434.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

technique(s)

dot blot: suitable
immunoprecipitation (IP): suitable
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys79)

Gene Information

human ... H3C1(8350)
mouse ... H3C1(360198)
rat ... H3C1(679994)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Predicted to react with multiple vertebrate species due to sequence conservation.
This antibody recognizes Histone H3 methylated at Lys79. Using peptide dot blot analysis slight cross-reactivity with dimethyl-Histone H3 (Lys79) and unmodified Histone H3 (Lys79) may be observed. Using peptide inhibition analysis assay no cross reactivity to these peptides is observed.

Immunogen

Epitope: Methylated at Lys79.
KLH-conjugated linear peptide corresponding to human Histone H3 monmethylated at Lys79.

Application

Dot Blot Analysis: A representative lot detected monomethyl-Histone H3 in Dot Blot.

Peptide Inhibition Analysis: A representative lot blocked monomethyl-Histone H3 in HeLa acid extract.

Immunoprecipitation Analysis: A representative lot detected monomethyl-Histone H3 in HeLa acid extract.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Use Anti-monomethyl-Histone H3 (Lys79) Antibody (Rabbit Polyclonal Antibody) validated in WB, DB, PIA, IP to detect monomethyl-Histone H3 (Lys79) also known as H3K79me1, Histone H3 (mono methyl K79).

Quality

Evaluated by Western Blot in HeLa acid extract.

Western Blot Analysis: A 1:500 dilution of this antibody detected monomethyl-Histone H3 in HeLa acid extract.

Target description

~17 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa acid extract.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muna M Hugais et al.
microPublication biology, 2021 (2021-11-09)
Alzheimer's Disease (AD), the most common type of dementia, is a neurodegenerative disease characterized by plaques of amyloid-beta (Aβ) peptides found in the cerebral cortex of the brain. The pathological mechanism by which Aβ aggregation leads to neurodegeneration remains unknown.
Samantha N Cobos et al.
Pathogens (Basel, Switzerland), 11(12) (2022-12-24)
Prions are proteins able to take on alternative conformations and propagate them in a self-templating process. In Saccharomyces cerevisiae, prions enable heritable responses to environmental conditions through bet-hedging mechanisms. Hence, [PRION+] states may serve as an atypical form of epigenetic
Karen Chen et al.
ACS chemical neuroscience, 9(4), 838-848 (2017-12-16)
Amyotrophic lateral sclerosis (ALS) and Parkinson's disease (PD) are devastating neurodegenerative diseases involving the progressive degeneration of neurons. No cure is available for patients diagnosed with these diseases. A prominent feature of both ALS and PD is the accumulation of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service