Skip to Content
Merck
All Photos(1)

Key Documents

ABE1314

Sigma-Aldrich

Anti-phospho TIN2 (Ser295) Antibody

0.25 mg/mL, from rabbit

Synonym(s):

TERF1-interacting nuclear factor 2, TRF1-interacting nuclear protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CHF 301.00

CHF 301.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
200 μL
CHF 301.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 301.00


Estimated to ship on23 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

concentration

0.25 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer295)

Gene Information

human ... TINF2(26277)

General description

The telomere-associated protein 1 interacting nuclear factor 2 (TIN2, TINF2, DKCA3) is a 40kDa protein critical to telomere function. TIN2 has no known enzymatic activity, but interacts with double-stranded and single-stranded telomeric DNA-binding proteins. Specifically, TIN2 directly binds to the telomeric dsDNA-binding proteins TRF1 and TRF2, and to TPP1, which forms a heterodimer with the telomeric ssDNA-binding protein POT1. TIN2 functions in telomere length as TIN2 knockdown results in moderate telomere elongation, as well as destabilization of TRF1 and TRF2. Complete knockout of TIN2 causes severe telomere dysfunction. The action of TIN2 is at least partially mediated by phosphorylation. TIN2 is phosphorylated at S295 and S330 amino acids. The level of phosphorylated TIN2 is up regulated during mitosis. The consequence of TIN2 phosphorylation during mitosis remains to be determined as initial results using non-phosphorylated mutants failed to show any differences between mutants and wildtype phenotypes. Phospho-TIN2 (S295) is created by phosphorylating TIN2 by the kinase RSK2.

Immunogen

Epitope: Phosphorylated S295
Linear peptide corresponding to human TIN2 .

Application

Anti-phospho TIN2 (Ser295) antibody is an antibody against phospho TIN2 (Ser295) for use in western blotting.
Western Blotting Analysis: A 1:500 dilution from a representative lot detected TIN2 in 10 µg of HeLa cell lysate transfected with TIN2 AA mutant and HeLa cell lysate transfected with TIN2 wild-type.
Western Blotting Analysis: A representative lot detected TIN2 in nocodazole treated HeLa cell lysate (Yang, S., and Counter, C. M., PLoS One. 8(8):e71697).

Quality

Evaluated by Western Blotting in Hela acid extract cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected TIN2 in 10 µg of Hela acid extract cell lysate.

Target description

~40 kDa observed. Uniprot describes 3 isoforms produced by alternative splicing at ~50 kDa (Isoform 1), ~39 kDa (Isoform 2), and ~15 kDa (Isoform 3). Uncharacterized band(s) may appear in some lysates.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service