Skip to Content
Merck
All Photos(1)

Key Documents

ABE1313

Sigma-Aldrich

Anti-SMARCD3 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

affinity purified immunoglobulin

Quality Level

clone

polyclonal

purified by

affinity chromatography

species reactivity

rabbit, mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

General description

KLH-conjugated linear peptide corresponding to the SWIB domain of human SMARCD3.

Specificity

Predicted to cross react to both isoforms of SMARCD3.

Application

Epitope: SWIB domain

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 5.0 µg/mL of this antibody detected SMARCD3 in 10 µg of human kidney tissue lysate.

Physical properties

~58 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Human, Mouse. Predicted to react with Rat, Bovine based on 100% sequence homology.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service