Skip to Content
Merck
All Photos(2)

Key Documents

ABD99

Sigma-Aldrich

Anti-Rex-1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger protein 42, Reduced expression protein 1, mREX-1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 311.00

CHF 311.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 311.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 311.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ZFP42(132625)

General description

The Rex-1 (Zfp-42) gene, which encodes an acidic zinc finger protein, is expressed at high levels in embryonic stem (ES) and F9 teratocarcinoma cells. The Rex-1 promoter is regulated by specific octamer family members in early embryonic cells, and is involved in retinoic acid (RA)-associated reduction in expression. Rex-1 is Involved in self-renewal property of ES cells and may be involved in transcriptional regulation of pluripotency signaling pathways. Rex-1 is also involved in the reprogramming of X-chromosome inactivation during the acquisition of pluripotency and is required for efficient elongation of TSIX, a non-coding RNA antisense to XIST.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of Human Rex-1.

Application

Detect Rex-1 using this Anti-Rex-1 Antibody validated for use in western blotting & ICC.
Immunocytochemsitry Analysis: A 1:100 dilution of this antibody detected Rex-1 in a undifferentiated H9 human embryonic stem cells.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Quality

Evaluated by Western Blotting in a undifferentiated H9 human embryonic stem cell lysate. Western Blotting Analysis: 1.0 µg/mL of this antibody detected Rex-1 in undifferentiated H9 human embryonic stem cells.

Target description

~35 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Undifferentiated H9 human embryonic stem cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service