Skip to Content
Merck
All Photos(1)

Key Documents

ABC822

Sigma-Aldrich

Anti-LIMP II Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Lysosome membrane protein 2, 85 kDa lysosomal membrane sialoglycoprotein, LGP85, CD36 antigen-like 2, Lysosome membrane protein II, LIMP II, Scavenger receptor class B member 2, CD36

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 352.00

CHF 352.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 352.00


Estimated to ship on29 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SCARB2(950)

General description

Lysosome membrane protein 2, also known as 85 kDa lysosomal membrane sialoglycoprotein (LGP85) or Lysosome membrane protein II (LIMP II) or Scavenger receptor class B member 2, and encoded by the gene name SCARB2 or CD36L2 or LIMPII, is a heavily glycosylated type III transmembrane protein, the majority of which exists in the lumen of the lysosome and a cytoplasmic domain of approximately 20 amino acids. A deficiency of LIMP II in mice causes uretic pelvic junction obstruction, deafness, and peripheral neuropathy associated with impaired vesicular trafficking and distribution of apically expressed proteins. More recently, LIMP II was shown to act as a receptor to bind β-glucocerebrosidase, the enzyme defective in Gaucher disease, a lysosomal storage disorder. LIMP II-deficient mice showed missorted as well as secreted β-glucocerebrosidase, suggesting that LIMP II also functions as the mannose-6-phosphate-independent trafficking receptor.

Immunogen

Epitope: center region
KLH-conjugated synthetic peptide corresponding to the center region of human LIMP II.

Application

Anti-LIMP II Antibody is an antibody against LIMP II for use in western blotting, IHC & immunofluorescence.
Immunohistochemistry Analysis: 2.5 μg/mL from a representative lot detected LIMP II in human skeletal muscle tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected LIMP II in human skeletal muscle cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in human skeletal muscle tissue lysate with and without blocking peptide.

Western Blotting Analysis: 1 μg/mL of this antibody detected GPAT-1 in human skeletal muscle tissue lysate with and without blocking peptide.

Target description

~80 kDa observed. Despite its predicted molecular weight, LIMP II runs at approximately 80 – 85 kDa in Western Blotting.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service