ABC474
Anti-Fhl1 Antibody
serum, from rabbit
Synonym(s):
Four and a half LIM domains protein 1, FHL-1, Skeletal muscle LIM-protein 1, SLIM, SLIM-1
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, human, rat
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FHL1(2273)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Fhl1 in an additional panel of select tissue lysates.
Western Blotting Analysis: A representative lot from an independent laboratory detected Fhl1 in nonanchored and anchored CasKO and Cx43KO cell lysates. A representative lot from the same independent laboratory detected Fhl1 in non-SRC transformed Cx43KO cell lysate and not in SRC transformed Cx43KO cell lysate (Shen, Y., et al. (2006). Cancer Res. 66(3):1543-1552.).
Western Blotting Analysis: A representative lot from an independent laboratory detected Fhl1 in non-SRC transformed Cx43KO cell lysate and not in SRC transformed Cx43KO cell lysate (Li, X., et al. (2008). Cancer Sci. 99(7):1326-1333.).
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Fhl1 in normal brain, kidney, liver, lung, prostate, and skin tissues and demonstrated a loss of signal in breast carcinoma, renal carcinoma, hepatocarcinoma, pulmonary carcinoma, prostatic carcinoma, and melanoma tissues (Shen, Y., et al. (2006). Cancer Res. 66(3):1543-1552.).
Immunohistochemistry Analysis: A representative lot from an independent laboratory detected Fhl1 in normal epithelium of breast, kidney, and prostate tissues, and demonstrated a loss of signal in mammary duct carcinoma, renal cell carcinoma, and prostate adenocarcinoma tissues (Li, X., et al. (2008). Cancer Sci. 99(7):1326-1333.).
Immunocytochemistry Analysis: A representative lot from an independent laboratory detected Fhl1 in non SRC transformed Cx43KO and CasKO cells and demonstrated a loss of signal in SRC transformed Cx43KO and CasKO cells (Shen, Y., et al. (2006). Cancer Res. 66(3):1543-1552.).
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Fhl1 in 10 µg of human brain tissue lysate.
Target description
Linkage
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service