Skip to Content
Merck
All Photos(2)

Key Documents

ABC327

Sigma-Aldrich

Anti-PD-L2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Programmed cell death 1 ligand 2, PD-1 ligand 2, PD-L2, PDCD1 ligand 2, Programmed death ligand 2, Butyrophilin B7-DC, B7-DC, CD273

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 371.00

CHF 371.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 371.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 371.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Programmed cell death ligand 2 (PD-L2, PD-1 ligand 2, PDCD1 ligand 2), also known as Butyrophilin B7-DC (B7-DC), is a member of the BTN/MOG family and immunoglobulin superfamily which is thought to act as a negative regulator of T cell activation. PD-L2 may play a role in stimulating an immunogenic response through an alternative receptor from PD-1. PD-L2 is highly expressed in heart, placenta, pancreas, lung, and liver and weakly expressed in spleen, lymph nodes, and thymus.

Specificity

This antibody only recognizes the largest isoform at ~31 kDa.

Immunogen

KLH-conjugated linear peptide corresponding to human PD-L2.

Application

Anti-PD-L2 Antibody is a highly specific rabbit polyclonal antibody, that targets Programmed cell death 1 ligand 1 & has been tested in western blotting, IHC (Paraffin) & Immunofluorescence.
Immunohistochemistry Analysis: 2.5 µg/mL from a representative lot detected PD-L2 in mouse brain tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected PD-L2 in mouse brain cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in Raji cell lysate

Western Blotting Analysis: 2 µg/mL of this antibody detected PD-L2 in 15 µg of Raji cell lysate.

Target description

~31 kDa observed. Uniprot describes 2 isoforms at ~21 kDa and one isoform at ~31 kDa Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Raji cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service