Skip to Content
Merck
All Photos(2)

Key Documents

ABC1392

Sigma-Aldrich

Anti-PDGF-C (GFD)

from rabbit

Synonym(s):

Platelet-derived growth factor C, Fallotein, Spinal cord-derived growth factor, SCDGF, VEGF-E

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 341.00

CHF 341.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 341.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 341.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

monkey (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PDGFC(56034)

General description

Platelet-derived growth factor C (UniProt: Q9NRA1; also known as PDGF-C, Fallotein, Spinal cord-derived growth factor, SCDGF, VEGF-E) is encoded by the PDGFC (also known as SCDGE, UNQ174/PRO200) gene (Gene ID: 56034) in human. PDGF-C is a disulfide-linked homodimeric growth factor that plays an essential role in the regulation of embryonic development, cell proliferation, cell migration, survival and chemotaxis. It also plays an important role in wound healing and angiogenesis. It also serves as a potent mitogen and chemoattractant for cells of mesenchymal origin. It is synthesized with a signal peptide (aa 1-22), which is subsequently cleaved off in the mature form. Its CUB domain is localized to amino acids 46 to 163 and proteolytic removal of the CUB domain is essential for unmasking the receptor-binding epitopes of the core domain. PDGF-C is expressed in the fallopian tube, vascular smooth muscle cells in kidney, breast and colon and in visceral smooth muscle of the gastrointestinal tract. Highly expressed in retinal pigment epithelia. Its high expression is also reported in retinal pigment epithelia. In the kidney, it is constitutively expressed in parietal epithelial cells of Bowman′s capsule, tubular epithelial cells, and in arterial endothelial cells. Higher levels of PDGF-C and have been reported in the malignant breast tumor microenvironment. In MCF-7 cells increased expression of PDGF-C is reported to enhance cell proliferation and tumor cell motility. Tissue plasminogen activator and matriptase are shown to process PDGF-C in MCF-7 cells to its active form. (Ref.: Hurst, NJ., et al. (2012). Biochem. J. 441(3); 909-918; Lei, H., and Kazlauskas, A (2008). Exp. Eye Res. 86(5); 711-712).

Specificity

This rabbit polyclonal antibody detects full length platelet derived growth factor C (PDGF-C) and also the 18 kDa growth factor domain.

Immunogen

A linear peptide corresponding to 21 amino acids from the C-terminal growth factor domain of human Platelet-derived growth factor C (PDGF-C).
Epitope: C-terminus

Application

Anti-PDGF-C, Cat. No. ABC1392, is a rabbit polyclonal antibody that detects Platelet-derived growth factor C and has been tested for use in Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected PDGF-C in Western Blotting applications (Hurst, N.J., et. al. (2012). Biochem J. 441(3):909-18).

Western Blotting Analysis: A 1:2,000 dilution from a representative lot detected PDGF-C in HEK293T cells. (Courtesy of Hyeong-Reh Choi Kim lab at Wayne State University).

Quality

Evaluated by Western Blotting in HEK293T cells.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected PDGF-C in lysates from HEK293T cells.

Target description

~48 kDa observed; 39.03 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Li et al.
Experimental and therapeutic medicine, 21(4), 289-289 (2021-03-16)
Renal interstitial fibrosis is the most important pathological process in chronic renal failure. Previous studies have shown that poricoic acid A (PAA), the main chemical constituent on the surface layer of the mushroom Poria cocos, has protective effects against oxidative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service