Skip to Content
Merck
All Photos(1)

Key Documents

AB9612

Sigma-Aldrich

Anti-Brain Derived Neurotrophic Factor Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CHF 331.00

CHF 331.00


Estimated to ship on11 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1136

Request a Bulk Order

Select a Size

Change View
50 μL
CHF 331.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 331.00


Estimated to ship on11 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1136

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... BDNF(627)

Specificity

Brain Derived Neurotrophic Factor (BDNF). By Western blot the antibody reacts with recombinant BDNF protein and also detects the ~28 kDa proBDNF protein in rat brain extracts. The antibody shows no cross reactivity with NGF, NT3 or NT4/5 by Western blot.

Immunogen

Synthetic peptide from mouse BDNF.

Application

Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain
This Anti-Brain Derived Neurotrophic Factor Antibody is validated for use in WB for the detection of Brain Derived Neurotrophic Factor.
Western blot: 1:500-1:2,000

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Lyophilized. Reconstitute with 50 μL of sterile distilled water. Centrifuge to remove any insoluble material.

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Romain Neidl et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 36(15), 4351-4361 (2016-04-15)
Aging weakens memory functions. Exposing healthy rodents or pathological rodent models to environmental enrichment (EE) housing improves their cognitive functions by changing neuronal levels of excitation, cellular signaling, and plasticity, notably in the hippocampus. At the molecular level, brain derived-neurotrophic
A novel activator of CBP/p300 acetyltransferases promotes neurogenesis and extends memory duration in adult mice.
Chatterjee, S; Mizar, P; Cassel, R; Neidl, R; Selvi, BR; Mohankrishna, DV; Vedamurthy et al.
The Journal of Neuroscience null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service