Skip to Content
Merck
All Photos(1)

Key Documents

AB5970

Sigma-Aldrich

Anti-Dishevelled-1 Antibody

Chemicon®, from rabbit

Synonym(s):

DSH, DVL1, DSH Homolog 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CHF 328.00

CHF 328.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
50 μL
CHF 328.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 328.00


Estimated to ship on14 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

input

sample type hematopoietic stem cell(s)
sample type neural stem cell(s)

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DVL1(1855)

General description

DVL1, the human homolog of the Drosophila dishevelled gene (dsh) encodes a cytoplasmic phosphoprotein that regulates cell proliferation. The protein acts as a transducer molecule for developmental processes, including segmentation and neuroblast specification. DVL1 mutation is likely responsible for transformation in neuroblastomas, and the Schwartz-Jampel syndrome and Charcot-Marie-Tooth disease type 2A have been mapped to the same region as DVL1 gene, thus DVL1 maybe important in these diseases as well.

Specificity

Dishevelled-1 protein (dvl-1). By Western blot the antibody shows no cross reactivity with recombinant dishevelled -2 or dishevelled -3 protein Pre-adsorption of the antibody with the immunogen peptide completely abolishes its reactivity by Western blot against dishevelled -1 protein.

Immunogen

Synthetic peptide corresponding to amino acids 557-571 of human dishevelled-1 protein.

Application

Anti-Dishevelled-1 Antibody is an antibody against Dishevelled-1 for use in WB.
Research Category
Stem Cell Research
Research Sub Category
Hematopoietic Stem Cells

Neural Stem Cells
Western blot: 1:500 using ECL. The antibody reacts with a protein of ~85kDa. Dilute antibody in 50 mM PBS, pH 7.4 containing 1.5% sodium chloride and 1% normal animal serum (same species as the host of the secondary antibody).

Optimal working dilutions must be determined by end user.

Target description

85 kDa

Physical form

Liquid in PBS, pH 7.4, containing 0.01M sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots (not less than 10 μL) for up to 6 months. Avoid repeated freeze/thaw cycles. The use of high quality antiserum-grade plastic or glass is recommended.

Analysis Note

Control
SK-N-SH (neuroblastoma) whole cell lysates

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micha Nethe et al.
Journal of cell science, 125(Pt 14), 3430-3442 (2012-04-03)
The Rho-GTPase Rac1 promotes actin polymerization and membrane protrusion that mediate initial contact and subsequent maturation of cell-cell junctions. Here we report that Rac1 associates with the ubiquitin-protein ligase neural precursor cell expressed developmentally down-regulated 4 (Nedd4). This interaction requires

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service