Skip to Content
Merck
All Photos(1)

Key Documents

AB5705

Sigma-Aldrich

Anti-Calcitonin Gene Related Protein Antibody

Chemicon®, from chicken

Synonym(s):

CGRP

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CHF 405.00

CHF 405.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
CHF 405.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 405.00


Estimated to ship on16 May 2025


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CALCA(796)

Specificity

CGRP (Calcitonin Gene Related Peptide).

Immunogen

A 14 amino acid synthetic peptide from rat CGRP I (processed active peptide). The immunogen sequence is identical at 13 of 14 residues between rat/mouse CGRP I and CGRP II. {KDNFVPTNVGSEAF-NH2}

Application

Immunohistochemistry: 1:500 See Protocol below.

Optimal working dilutions must be determined by the end user.



AB5705 has been used successfully on perfusion fixed tissue. Perfusion: 1) Ca2+-free Tyrode′s solution followed by 2) a paraformaldehyde-picric acid fixative and 3) 10% sucrose in PBS as a cryo-protectant. Tissues were rapidly dissected out and stored overnight in 0.1 M phosphate buffer (pH 7.4) containing 10% sucrose.Slide-mounted tissue sections were incubated with blocking buffer for 1 hour at room temperature. Primary antibody was diluted in blocking buffer to the appropriate working dilution. Blocking buffer was removed and the slides were then incubated at 2-8°C for 18-24 hours with AB5705. After rinsing in PBS 3 times sections were incubated for 60 minutes at room temperature with Cy3-conjugated secondary antibodies. After mounting in a mixture of PBS and glycerol (1:3) containing 0.1% p-phenylenediamine, sections were examined with a Nikon Microphot-SA epifluorescence microscope.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors
This Anti-Calcitonin Gene Related Protein Antibody is validated for use in IH for the detection of Calcitonin Gene Related Protein.

Physical form

Format: Purified
Purified chicken IgY. Liquid containing 0.05% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin J O'Donovan et al.
The Journal of experimental medicine, 211(5), 801-814 (2014-04-16)
Activation of intrinsic growth programs that promote developmental axon growth may also facilitate axon regeneration in injured adult neurons. Here, we demonstrate that conditional activation of B-RAF kinase alone in mouse embryonic neurons is sufficient to drive the growth of
Alessandro Faroni et al.
Molecular neurobiology, 56(2), 1461-1474 (2018-06-28)
GABA-B receptors are important for Schwann cell (SC) commitment to a non-myelinating phenotype during development. However, the P0-GABA-B1fl/fl conditional knockout mice, lacking the GABA-B1 receptor specifically in SCs, also presented axon modifications, suggesting SC non-autonomous effects through the neuronal compartment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service