Skip to Content
Merck
All Photos(1)

Key Documents

AB5350

Sigma-Aldrich

Anti-Potassium Channel SK3 Antibody

Chemicon®, from rabbit

Synonym(s):

KCa3, Kcnn3, SKCa3

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
CHF 868.00

CHF 868.00


Estimated to ship on16 April 2025


Request a Bulk Order

Select a Size

Change View
200 μL
CHF 868.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 868.00


Estimated to ship on16 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KCNN4(3783)

Specificity

Recognizes SK3. The epitope specific for SK3 is not present in any other known proteins. The immunogen sequence is identical in mouse and porcine.

Immunogen

Highly purified peptide corresponding to residues 2-21 of human SK3 (Accession Q9UGI6).

Application

Anti-Potassium Channel SK3 Antibody detects level of Potassium Channel SK3 & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blotting: 1.0-1.5 μg/mL (1:200-1:300)

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, 5% sucrose and 0.025% sodium azide. Reconstitute with 200 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 20 μg of control antigen (lyophilized powder). Reconstitute with 100 μL of sterile distilled water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam S Deardorff et al.
The Journal of physiology, 599(5), 1391-1420 (2021-01-16)
The spatial and temporal balance of spinal α-motoneuron (αMN) intrinsic membrane conductances underlies the neural output of the final common pathway for motor commands. Although the complete set and precise localization of αMN K+ channels and their respective outward conductances
Histone deacetylase inhibitors (HDACi) increase expression of KCa2.3 (SK3) in primary microvascular endothelial cells.
Kolski-Andreaco, et al.
American Journal of Physiology. Cell Physiology, 322, C338-C353 (2023)
S Shekar Dukkipati et al.
The Journal of physiology, 596(9), 1723-1745 (2018-03-05)
Motoneuron soma size is a largely plastic property that is altered during amyotrophic lateral sclerosis (ALS) progression. We report evidence of systematic spinal motoneuron soma size plasticity in mutant SOD1-G93A mice at various disease stages and across sexes, spinal regions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service