Skip to Content
Merck
All Photos(2)

Key Documents

AB5226

Sigma-Aldrich

Anti-P2X4 Receptor Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
CHF 346.00

CHF 346.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
50 μL
CHF 346.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 346.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

Recognizes a full-length P2X4 purinergic receptor protein and not with other known proteins tested so far.

SPECIES REACTIVITIES: The immunogen sequence is identical in mouse. Other species have not been tested.

Immunogen

A purified peptide of rat P2X4 protein (amino acids 370-388) (Accession P51577).

Application

Anti-P2X4 Receptor Antibody detects level of P2X4 Receptor & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1-1.5 μg/mL (1:200:1:300) using ECL

Immunohistochemistry on rat brain sections.

Dilutions should be made using a carrier protein such as BSA (1-3%)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
CONTROL ANTIGEN: Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of purified peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sobia Manzoor et al.
PloS one, 17(5), e0259727-e0259727 (2022-05-21)
Major HCV infections lead to chronic hepatitis, which results in progressive liver disease including fibrosis, cirrhosis and eventually hepatocellular carcinoma (HCC). P2X4 and P2X7 are most widely distributed receptors on hepatocytes. Full length P2X4 (1.7kb) (Rattus norvegicus) was sub cloned
Gaurav Datta et al.
Frontiers in cellular neuroscience, 13, 329-329 (2019-08-06)
Human immunodeficiency virus type 1 (HIV-1) associated neuropathy is the most common neurological complication of HIV-1, with debilitating pain affecting the quality of life. HIV-1 gp120 plays an important role in the pathogenesis of HIV neuropathy via direct neurotoxic effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service