Skip to Content
Merck
All Photos(1)

Key Documents

AB3572P

Sigma-Aldrich

Anti-Organic Anion Transporting Polypeptide 2 Antibody, CT, intracellular

Chemicon®, from rabbit

Synonym(s):

oatp2, OATP-B1, Brain digoxin carrier protein

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
CHF 472.00

CHF 472.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
CHF 472.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 472.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

General description

Organic Anion Transporting Polypeptide 2 is an integral membrane protein that mediates the sodium-independent transport of organic ions, cardiac glycosides and thyroid hormones. Oatp2 is highly expressed in brain, liver, and kidney but not expressed in heart, spleen, lung, skeletal muscle, or testis.

Specificity

Recognizes rat Organic Ion Transporter 2 Polypeptide 2.

Immunogen

A 12 amino acid peptide sequence near the cytoplasmic, C-terminus of rat OATP2. The immunogen shows no significant sequence similarity to other OATPs or other proteins.

Application

Detect Organic Anion Transporting Polypeptide 2 using this Anti-Organic Anion Transporting Polypeptide 2 Antibody, C-terminus, intracellular validated for use in ELISA & WB.
Western blot: 1-10 μg/mL using ECL. Rat OATP2 is approximately 75 kD in brain and liver (Noe et al. 1997).

ELISA: 0.5-1.0 μg/mL using 1 μg/mL control peptide per well.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS, containing 0.1% BSA.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Camilla A Thorling et al.
Journal of biomedical optics, 18(10), 101306-101306 (2013-07-03)
The liver is important in the biotransformation of various drugs, where hepatic transporters facilitate uptake and excretion. Ischemia-reperfusion (I/R) injury is a common occurrence in liver surgery, and the developing oxidative stress can lead to graft failure. We used intravital

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service