Skip to Content
Merck
All Photos(1)

Key Documents

AB3517

Sigma-Aldrich

Anti-Polyhistidine Tag Antibody

Chemicon®, from chicken

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 346.00

CHF 346.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 346.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

CHF 346.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Polyhistidine tag, 6-His Tag containing proteins

Immunogen

Synthetic peptide consisting of six histidines.

Application

Anti-Polyhistidine Tag Antibody detects level of Polyhistidine Tag & has been published & validated for use in ELISA, WB & IC.
Immunoblotting: 1:250-1:2,000

Immunocytochemistry: 1:250-1:1,000

ELISA: 1:250-1:2,000

Optimal working dilutions must be determined by the end user.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rebba C Boswell-Casteel et al.
Molecules (Basel, Switzerland), 23(4) (2018-03-23)
Equilibrative nucleoside transporters (ENTs) are polytopic membrane transporters responsible for the translocation of nucleosides, nucleobases-to a lesser extent-and nucleoside analog therapeutics across cellular membranes. ENTs function in a diffusion controlled bidirectional manner and are thought to utilize an alternating access

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service