Skip to Content
Merck
All Photos(1)

Key Documents

AB3421

Sigma-Aldrich

Anti-Complement C3 Antibody

Chemicon®, from chicken

Synonym(s):

Anti-AHUS5, Anti-ARMD9, Anti-ASP, Anti-C3a, Anti-C3b, Anti-CPAMD1, Anti-HEL-S-62p

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CHF 639.00

CHF 639.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
CHF 639.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 639.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... C3(718)

Specificity

Complement C3

Immunogen

Human C3, full length plasma protein.

Application

Detect Complement C3 using this Anti-Complement C3 Antibody validated for use in ELISA.
ELISA, direct: 0.1 μg/mL

It is expected that the antibody will also work for Western blot, immunohistochemistry and immunocytochemistry. Specific reactivity has not been confirmed.

Optimal working dilutions must be determined by the end user.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Chicken IgY fraction, in 0.15 M NaCl, 0.02 M NaH2PO4, 0.1% NaN3, pH 7.2.

Storage and Stability

Maintain at 2-8*C in undiluted aliquots up to 6 months after date of receipt.
Avoid freezing temperatures.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pizga Kumwenda et al.
Cell reports, 38(1), 110183-110183 (2022-01-06)
Candida albicans is a commensal of the urogenital tract and the predominant cause of vulvovaginal candidiasis (VVC). Factors that increase circulatory estrogen levels such as pregnancy, the use of oral contraceptives, and hormone replacement therapy predispose women to VVC, but
Kelsey B Law et al.
Autophagy, 13(5), 868-884 (2017-05-20)
Peroxisome biogenesis disorders (PBDs) are metabolic disorders caused by the loss of peroxisomes. The majority of PBDs result from mutation in one of 3 genes that encode for the peroxisomal AAA ATPase complex (AAA-complex) required for cycling PEX5 for peroxisomal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service