Skip to Content
Merck
All Photos(1)

Key Documents

AB3241

Sigma-Aldrich

Anti-Cdc2 Antibody, phospho-specific (Tyr15)

Chemicon®, from rabbit

Synonym(s):

Cell Division Cycle 2, p34 Protein Kinase, Cyclin-dependent Kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr15)

Gene Information

rat ... Cdk1(54237)

Specificity

CDC2, phosphoTyr15. By Western blot the antibody recognizes the ~35 kDa phospho CDC2 protein. The labeling of the CDC2 band in Western blots of rat brain is blocked by the phosphopeptide but not by the dephosphopeptide.

Immunogen

Epitope: phospho-specific (Tyr15)
Synthetic peptide corresponding to amino acids surrounding the phosphoTry15 of human, rat, mouse and Xenopus CDC2.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-Cdc2 Antibody, phospho-specific (Tyr15) (Rabbit Polyclonal Antibody) validated in WB to detect Cdc2 also known as Cell Division Cycle 2, p34 Protein Kinase.
Western blot: 1:1,000

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin in 10 mM Hepes, pH 7.5, with 150 mM NaCl, 100 μg/mL BSA and 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted for up to 6 months from date of receipt. Avoid repeated freeze/thaw cycles. Do not store in a self-defrosting freezer.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service