Skip to Content
Merck
All Photos(2)

Key Documents

AB1781SP

Sigma-Aldrich

Anti-Neurotrophin 4/5 Antibody

Chemicon®, from rabbit

Synonym(s):

NT4/5

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
CHF 352.00

CHF 352.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
CHF 352.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 352.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, monkey, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NTF4(4909)

Specificity

Neurotrophin 4 (NT4). By dot blot, less than 1% cross-reactivity against NGF, BDNF or NT3.

Immunogen

Recombinant human Neurotrophin 4 (NT4).

Application

Anti-Neurotrophin 4/5 Antibody is an antibody against Neurotrophin 4/5 for use in ELISA, WB, IH.
Immunohistochemistry: 1-2 μg/mL (see suggested protocol).

Immunoblotting: 1-2 μg/mL

ELISA: 1-2 μg/mL

Inhibition of biological activity in vitro: 1-2 μg/mL.
BIOLOGICAL ACTIVITY: Neutralizes NT4, but not other neurotrophins.

In vivo animal studies: use neat

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Linkage

Replaces: AB1455P

Physical form

Format: Purified
Purified immunoglobulin. Lyophilized from PBS, no preservatives. Reconstitute with 500 μL of sterile distilled water (1 mg/mL).

Storage and Stability

Maintain lyophilized material at -20 to -70°C for up to 12 months after date of receipt. After reconstitution maintain at -20 to -70°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Cabo et al.
Anatomical record (Hoboken, N.J. : 2007), 298(3), 624-629 (2014-09-19)
Some mechanoreceptors in mammals depend totally or in part on the neurotrophins brain-derived neurotrophic factor (BDNF) and neurotrophin-4 (NT-4), and their receptor TrkB, for development and maintenance. These actions are presumably exerced regulating the survival of discrete sensory neurons in
Takahiro Ishimoto et al.
NPJ science of food, 8(1), 11-11 (2024-02-07)
Oral administration of the food-derived antioxidant amino acid ergothioneine (ERGO) results in its efficient distribution in the brain and enhances cognitive function. However, effect of ERGO deficiency on cognitive impairment and the underlying mechanisms remain unknown. We revealed that cognitive
Erica Hurtado et al.
Frontiers in molecular neuroscience, 10, 147-147 (2017-06-03)
The neurotrophin brain-derived neurotrophic factor (BDNF) acts via tropomyosin-related kinase B receptor (TrkB) to regulate synapse maintenance and function in the neuromuscular system. The potentiation of acetylcholine (ACh) release by BDNF requires TrkB phosphorylation and Protein Kinase C (PKC) activation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service