Skip to Content
Merck
All Photos(1)

Key Documents

AB1592P

Sigma-Aldrich

Anti-NMDAR2C Antibody, also recognizes NMDAR2A and 2B

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
CHF 415.00

CHF 415.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 μG
CHF 415.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 415.00


Estimated to ship on26 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

suitability

not suitable for immunohistochemistry

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN2C(2905)

Specificity

NMDAR2C. Immunolabels the 140 kD NMDAR2C subunit of the NMDA receptor. Also reacts with NMDAR2A and NMDAR2B by Western blot (180 kD).

Immunogen

Epitope: N-terminus
N-Terminal Fusion Protein of the NMDAR2C

Application

Detect NMDAR2C using this Anti-NMDAR2C Antibody, also recognizes NMDAR2A & 2B validated for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blotting 1:100-1:200

Immunoprecipitation: 3 μL will (under the appropriate conditions) quantitatively immunoprecipitate all NMDAR2C in 200 μg of rat cerebellum.

Not recommended for use in immunohistochemistry since the antibody also reacts with NMDAR2A and NMDAR2B.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from ammonium bicarbonate (5 mM), some residual salt may be present. Reconstitute with 50 μL of PBS.

Storage and Stability

Maintain lyophilized antibody at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to six months.

Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josette S Lindahl et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 29(11), 2065-2073 (2004-05-13)
Phencyclidine (PCP) is a noncompetitive antagonist of the N-methyl-D-aspartate (NMDA) glutamate receptor subtype. It produces transient psychoses in normal individuals and exacerbates psychoses in schizophrenics. When administered to rodents, PCP elicits stereotypic behaviors including unrelenting head swaying, hyperlocomotion, and social
Ziad Hassan et al.
Cells, 12(9) (2023-05-13)
Impairment of one-carbon metabolism during pregnancy, either due to nutritional deficiencies in B9 or B12 vitamins or caused by specific genetic defects, is often associated with neurological defects, including cognitive dysfunction that persists even after vitamin supplementation. Animal nutritional models

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service