Skip to Content
Merck
All Photos(1)

Key Documents

AB1224

Sigma-Aldrich

Anti-Myeloperoxidase Antibody, human white cell

serum, Chemicon®

Synonym(s):

MPO

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
CHF 592.00

CHF 592.00


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
2 ML
CHF 592.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CHF 592.00


Estimated to ship on15 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Myeloperoxidase (Human White Cells). Monospecific by IEP. Some cross reactivity with Myeloperoxidase from other tissues and species may occur.

Myeloperoxidase (MPO) represents an early appearing and highly reliable intracellular myeloid lineage marker molecule. MPO protein can be detected in a subset of human hematopoietic bone marrow progenitor cells and in granulomonopoietic (GM) cells. However, other myeloid-related cell types such as epidermal Langerhans-type dendritic cells (LC) lack MPO. Therefore, human myeloid progenitors might be subdivided based on MPO protein expression into functional subsets.

Immunogen

Epitope: human white cell
Myeloperoxidase (Human White Cells)

Application

Anti-Myeloperoxidase Antibody, human white cell is an antibody against Myeloperoxidase for use in ELISA & WB.
Immunoblotting: antibody recognizes alpha and beta subunits of MPO. Reduced and denatured extracts of purified MPO produce blots produce 55-60kDa alpha subunits and 12-14kDa beta subunit bands140 Additional bands may be present due to cross reactivity with other myeloperoxidases.

ELISA:

Optimal working dilutions must be determined by end user.

Physical form

Monospecific antiserum purified by delipidation and defibrination. Lyophilized from 0.02 M Potassium Phosphate buffer, 0.15 M Sodium Chloride, pH 7.2, 0.01% Sodium Azide. Reconstitute with 2.0 mL of sterile distilled water.

Storage and Stability

Store unopened vial at 2-8 ºC. Maintain frozen at -20ºC in undiluted aliquots for up to one year after reconstitution. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service