Skip to Content
Merck
All Photos(1)

Key Documents

902262

Supelco

Coupling part

10 mm connector, tubing for 6x8 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
CHF 134.00

CHF 134.00


Estimated to ship on11 May 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
CHF 134.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

CHF 134.00


Estimated to ship on11 May 2025


Request a Bulk Order

fitting

10 mm connector
tubing for 6x8 mm

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

Our withdrawal and safety components are easily interconnectable thanks to a wide selection of reducers, adapters and couplings. Due to the wide variety, many different customized installations are possible – everything for your individual requirements.
This rapid-action connector is made of stainless steel with a special high quality sealing for maximum protection of our solvents quality. It is equipped with pipe connector for 10 mm pipes and tube connector 8x6 mm.
between tube (6x8 mm) and pipe (O.D. 10 mm)

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service