Skip to Content
Merck
All Photos(1)

Key Documents

8.52309

Sigma-Aldrich

Fmoc-ACA-OH

Novabiochem®

Synonym(s):

Fmoc-ACA-OH, 7-N-Fmoc-aminocoumarin-4-acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
CHF 323.00
1 G
CHF 413.00

CHF 323.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
250 MG
CHF 323.00
1 G
CHF 413.00

About This Item

Empirical Formula (Hill Notation):
C26H19NO6
CAS Number:
Molecular Weight:
441.43
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

CHF 323.00


0 left in stock

Request a Bulk Order

Quality Level

product line

Novabiochem®

Assay

≥95.0% (HPLC)
≥98% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

aromatic amine

storage temp.

2-8°C

SMILES string

N(c4cc5[o][c](cc(c5cc4)CC(=O)O)=O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C26H19NO6/c28-24(29)11-15-12-25(30)33-23-13-16(9-10-17(15)23)27-26(31)32-14-22-20-7-3-1-5-18(20)19-6-2-4-8-21(19)22/h1-10,12-13,22H,11,14H2,(H,27,31)(H,28,29)

InChI key

RCUKLQDGAASIAX-UHFFFAOYSA-N

General description

Fmoc-ACA-OH is a useful tool for the SPPS of AMC fluorogenic protease substrates by Fmoc SPPS. Fmoc-ACA-OH [1] must be first loaded onto a Wang-type resin. Any unreacted linker hydroxyls must be capped using acetic anhydride. Following Fmoc removal, the first amino acid should be coupled with HATU/collidine [2]. Insertion of ACA into the middle of a peptide provides fluorogenic substrates for endopeptidases [3]. Treatment of the ACC peptide obtained after TFA cleavage with an aqueous base causes facile decarboxylation and formation of the AMC peptide.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Chromogenic Labeling of Synthetic Peptides

Literature references

[1] J. L. Harris, et al. (2000) Proc. Natl. Acad. Sci. USA, 97, 7754.
[2] D. J. Maly, et al. (2002) J. Org. Chem., 67, 910.
[3] C. M. Salisbury, et al. (2002) J. Am. Chem. Soc., 124, 14868.

Analysis Note

Color (visual): white to slight yellow to light red
Appearance of substance (visual): powder
Identity (IR): passes test
Purity (TLC(CMA1)): ≥ 98 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service