Skip to Content
Merck
All Photos(1)

Key Documents

8.44000

Sigma-Aldrich

Dibasic ester

(mixture of dimethyladipate, dimethylglutarate, dimethylsuccinate) for synthesis

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CHF 27.90

CHF 27.90


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
CHF 27.90

About This Item

UNSPSC Code:
12352108
NACRES:
NA.22

CHF 27.90


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

form

liquid

density

1.09 g/cm3 at 20 °C

storage temp.

2-30°C

General description

(mixture of dimethyladipate, dimethylglutarate, dimethylsuccinate) for synthesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service