Skip to Content
Merck
All Photos(1)

Key Documents

8.43788

Sigma-Aldrich

1,8-Cineole

for synthesis

Synonym(s):

1,8-Cineole, 1,3,3-Trimethyl-2-oxabicyclo(2,2,2)-octane

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CHF 42.60
500 ML
CHF 164.00

CHF 42.60


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 ML
CHF 42.60
500 ML
CHF 164.00

About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352005
EC Index Number:
207-431-5
NACRES:
NA.22

CHF 42.60


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2480 mg/kg LD50, oral (Rat)

mp

1.5 °C

transition temp

flash point 49 °C

solubility

3.5 g/L

density

0.925 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O1[C@@]2(CC[C@H](C1(C)C)CC2)C

InChI

1S/C10H18O/c1-9(2)8-4-6-10(3,11-9)7-5-8/h8H,4-7H2,1-3H3/t8-,10+

InChI key

WEEGYLXZBRQIMU-WAAGHKOSSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.923 - 0.928
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service