Skip to Content
Merck
All Photos(1)

Key Documents

8.43377

Sigma-Aldrich

DL-Tartaric acid

for synthesis

Synonym(s):

DL-Tartaric acid

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CHF 35.90
500 G
CHF 151.00

CHF 35.90


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
CHF 35.90
500 G
CHF 151.00

About This Item

Empirical Formula (Hill Notation):
C4H6O6
CAS Number:
Molecular Weight:
150.09
MDL number:
UNSPSC Code:
12352101
EC Index Number:
205-105-7
NACRES:
NA.22

CHF 35.90


Estimated to ship on01 June 2025


Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

autoignition temp.

425 °C

pH

1.6 (25 °C, 100 g/L in H2O)

mp

198-204 °C

transition temp

flash point 210 °C

solubility

215 g/L

storage temp.

2-30°C

SMILES string

[O-]C(=O)C(O)C(O)C(=O)[O-].[H+].[H+]

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)

InChI key

FEWJPZIEWOKRBE-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Melting range (lower value): ≥ 198 °C
Melting range (upper value): ≤ 204 °C
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service